6-Bromo-2,2'-bipyridyl 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 6-Bromo-2,2'-bipyridine

OR53209-1G OR53209-5G
APOSOR53209-1GEA 67.6 GBP
APOSOR53209-1G APOSOR53209-5G
6-Bromo-2,2'-bipyridyl 97%
6-Bromo-2,2'-bipyridyl
Formula: C₁₀H₇BrN₂
MW: 235.0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318953
CAS Number: 10495-73-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR