Dibenzylamine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR5296-1KG OR5296-250G
APOSOR5296-1KGEA 158 GBP
APOSOR5296-1KG APOSOR5296-250G
Dibenzylamine 98%
Dibenzylamine
Formula: C₆H₅CH₂NHCH₂C₆H₅
Boiling Pt: 300 °C
Melting Pt: –26 °C
Density: 1.026 g/cm³ (25 °C)
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00004770
CAS Number: 103-49-1
EINECS: 203-117-7
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR