5-Acetylvaleric acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 6-Oxoheptanoic acid

OR5276-10G
APOSOR5276-10GEA 26.3 GBP
APOSOR5276-10G
5-Acetylvaleric acid 98%
5-Acetylvaleric acid
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 158…162 °C/9mmHg
Melting Pt: 35…37 °C
Density: 1.059 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00051479
CAS Number: 3128-07-2
EINECS: 221-512-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR