(±)-1,2-Propanediol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Propylene glycol(±)-1,2-Propylene glycol1,2-Dihydroxypropane1,2-Propanediol(±)-1,2-Dihydroxy propane

APOSOR5249-500GEA 63.1 GBP
APOSOR5249-500G APOSOR5249-2.5KG
(±)-1,2-Propanediol 98%
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187 °C
Melting Pt: –60 °C
Density: 1.036 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR