2-Naphthalenemethanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR52473-100G OR52473-25G OR52473-10G
APOSOR52473-100GEA 101 GBP
APOSOR52473-100G APOSOR52473-25G APOSOR52473-10G
2-Naphthalenemethanol
2-Naphthalenemethanol
Formula: C₁₁H₁₀O
MW: 158.2000 g/mol
Boiling Pt: 178 °C/7.5mmHg
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00004124
CAS Number: 1592-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR