N-Methylmethanesulphonamide 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR52265-100G OR52265-25G
APOSOR52265-100GEA 105 GBP
APOSOR52265-100G APOSOR52265-25G
N-Methylmethanesulphonamide 97%
N-Methylmethanesulphonamide
Formula: C₂H₇NO₂S
Boiling Pt: 118 °C/0.3mmHg
Density: 1.28 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00776277
CAS Number: 1184-85-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR