1,5-Diphenylcarbazone 95%, Technical Grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: sym-Diphenylcarbazone, Phenylazoameisensäure-2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazide

OR52237-5G
APOSOR52237-5GEA 42 GBP
APOSOR52237-5G
1,5-Diphenylcarbazone 95%, Technical Grade
1,5-Diphenylcarbazone
Formula: C₁₃H₁₂N₄O
MW: 240.26 g/mol
Melting Pt: 154…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00003024
CAS Number: 538-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR