1,4-Bis(tributylstannyl)benzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR52022-1G OR52022-5G
APOSOR52022-1GEA 78.2 GBP
APOSOR52022-1G APOSOR52022-5G
1,4-Bis(tributylstannyl)benzene
1,4-Bis(tributylstannyl)benzene
Formula: C₃₀H₅₈Sn₂
Boiling Pt: 206…210 °C/0.04mmHg
Density: 1.147 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD17015284
CAS Number: 17151-51-8
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR