Ethyl-8-bromooctanoate 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR50964-100G
OR50964-25G
APOSOR50964-100GEA
111
GBP
APOSOR50964-100G
APOSOR50964-25G
Ethyl-8-bromooctanoate 97%
Ethyl-8-bromooctanoate
Formula:
C₁₀H₁₉BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00045057 CAS Number: 29823-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...