Denatonium benzoate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate

OR50488-100G OR50488-500G
APOSOR50488-100GEA 67.6 GBP
APOSOR50488-100G APOSOR50488-500G
Denatonium benzoate 98%
Denatonium benzoate
Formula: C₂₈H₃₄N₂O₃
MW: 446.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031578
CAS Number: 3734-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR