Maleic hydrazide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,2-Dihydro-3,6-pyridazinedione, 3,6-Dihydroxypyridazine, 3,6-Pyridazinediol, Maleic acid Hydrazide

OR4916-100G OR4916-25G OR4916-5G
APOSOR4916-100GEA 68.6 GBP
APOSOR4916-100G APOSOR4916-25G APOSOR4916-5G
Maleic hydrazide
Maleic hydrazide
Formula: C₄H₄N₂O₂
Melting Pt: 299…301 °C (dec.)
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006665
CAS Number: 123-33-1
EINECS: 204-619-9
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR