4'-Aminobenzanilide 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4′-Aminobenzanilide, 4-Aminobenzanilide

OR480682-25G
APOSOR480682-25GEA 168 GBP
APOSOR480682-25G
4'-Aminobenzanilide 95%
4'-Aminobenzanilide
Formula: C₁₃H₁₂N₂O
MW: 212.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035777
CAS Number: 17625-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR