Docosylamine
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR470890-1G
OR470890-250MG
OR470890-5G
APOSOR470890-1GEA
223
GBP
APOSOR470890-1G
APOSOR470890-250MG
APOSOR470890-5G
Docosylamine
Docosylamine
Formula:
H₃C(CH₂)₂₀CH₂NH₂ MW: 325.62 g/mol Storage Temperature: Ambient |
CAS Number:
14130-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...