4-Chloro-2-iodotoluene 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR460029-100G OR460029-50G
APOSOR460029-100GEA 57 GBP
APOSOR460029-100G APOSOR460029-50G
4-Chloro-2-iodotoluene 98%
4-Chloro-2-iodotoluene
Formula: C₇H₆ClI
Boiling Pt: 138 °C
Density: 1.82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00060661
CAS Number: 33184-48-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR