Lithiumbis(trimethylsilyl)amide 1 M in THF

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR460028-100ML OR460028-500ML
APOSOR460028-100MLEA 68.6 GBP
APOSOR460028-100ML APOSOR460028-500ML
Lithiumbis(trimethylsilyl)amide 1 M in THF
Lithium bis(trimethylsilyl)amide
Formula: C₆H₁₈LiNSi₂
Density: 0.891 g/cm³ (25 °C)
Flash Pt: 1.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR