3-Morpholin-4-ylpropionic acid hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR460024-5G OR460024-1G OR460024-250MG
APOSOR460024-5GEA 31.8 GBP
APOSOR460024-5G APOSOR460024-1G APOSOR460024-250MG
3-Morpholin-4-ylpropionic acid hydrochloride
3-Morpholin-4-ylpropionic acid hydrochloride
Formula: C₇H₁₄ClNO₃
Melting Pt: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00810444
CAS Number: 6319-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR