Isoamyl alcohol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR460021-250ML OR460021-500ML
APOSOR460021-250MLEA 68.6 GBP
APOSOR460021-250ML APOSOR460021-500ML
Isoamyl alcohol
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
Boiling Pt: 130 °C
Melting Pt: –117 °C
Density: 0.809 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR