N-Benzylphenethylamine 99%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OR451330-100G
OR451330-250G
OR451330-25G
APOSOR451330-100GEA
221
GBP
APOSOR451330-100G
APOSOR451330-250G
APOSOR451330-25G
N-Benzylphenethylamine 99%
N-Benzylphenethylamine
Formula:
C₁₅H₁₇N MW: 211.31 g/mol Boiling Pt: 162…165 °C Density: 1.007 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009624 CAS Number: 3647-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...