Ethyl ethoxyiminoacetate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR450017-1G OR450017-5G OR450017-500MG
APOSOR450017-1GEA 78.7 GBP
APOSOR450017-1G APOSOR450017-5G APOSOR450017-500MG
Ethyl ethoxyiminoacetate 95%
Ethyl ethoxyiminoacetate
Formula: C₆H₁₁NO₃
Density: 1.042 g/cm³ (25 °C)
Flash Pt: 72.4 °C
Storage Temperature: Freezer
MDL Number: MFCD06797654
CAS Number: 816-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR