1-Methyl-5-(tributylstannyl)imidazole, Technical Grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR4409-1G OR4409-5G
APOSOR4409-1GEA 105 GBP
APOSOR4409-1G APOSOR4409-5G
1-Methyl-5-(tributylstannyl)imidazole, Technical Grade
1-Methyl-5-(tributylstannyl)imidazole
Formula: C₁₆H₃₂N₂Sn
Boiling Pt: 160…164 °C/2mmHg
Density: 1.22 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319031
CAS Number: 147716-03-8
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR