3-Bromo-o-toluonitrile 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR43660-25G
APOSOR43660-25GEA 88.1 GBP
APOSOR43660-25G
3-Bromo-o-toluonitrile 98%
3-Bromo-o-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 44…45 °C
Storage Temperature: Ambient
MDL Number: MFCD09025665
CAS Number: 52780-15-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR