Bromotrichloromethane

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR43546-25G OR43546-100G
APOSOR43546-25GEA 26.3 GBP
APOSOR43546-25G APOSOR43546-100G
Bromotrichloromethane
Bromotrichloromethane
Formula: BrCl₃C
MW: 198.27 g/mol
Boiling Pt: 104…105 °C
Melting Pt: –6 °C
Density: 2.015 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000783
CAS Number: 75-62-7
EINECS: 200-886-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR