Aniline 99+% ACS

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Aminobenzene, Phenylamine

OR43506-100ML OR43506-1L OR43506-500ML
APOSOR43506-100MLEA 68.3 GBP
APOSOR43506-100ML APOSOR43506-1L APOSOR43506-500ML
Aniline 99+% ACS
Aniline
Formula: C₆H₅NH₂
Boiling Pt: 184 °C
Melting Pt: –6 °C
Density: 1.022 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007629
CAS Number: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR