Pyridine 99,8%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR41162-1L OR41162-500ML OR41162-5L
APOSOR41162-1LEA 31.5 GBP
APOSOR41162-1L APOSOR41162-500ML APOSOR41162-5L
Pyridine 99,8%
Pyridine
Formula: C₅H₅N
Boiling Pt: 115…116 °C
Melting Pt: –42 °C
Density: 0.978 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR