2,6-Di-tert-butyl-p-cresol 99%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
OR40350-1KG
OR40350-250G
OR40350-50G
APOSOR40350-1KGEA
89.8
GBP
APOSOR40350-1KG
APOSOR40350-250G
APOSOR40350-50G
2,6-Di-tert-butyl-p-cresol 99%
2,6-Di-tert-butyl-p-cresol
Formula:
C₁₅H₂₄O MW: 220.35 g/mol Boiling Pt: 264…265 °C Melting Pt: 68…73 °C Density: 1.048 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011644 CAS Number: 128-37-0 EINECS: 204-881-4 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...