Ethyl-3-[(2-furylmethyl)amino]propanoate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR3932-1G OR3932-5G
APOSOR3932-1GEA 160 GBP
APOSOR3932-1G APOSOR3932-5G
Ethyl-3-[(2-furylmethyl)amino]propanoate 97%
Ethyl-3-[(2-furylmethyl)amino]propanoate
Formula: C₁₀H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00219838
CAS Number: 175203-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR