4-(Tributylstannyl)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR3780-1G OR3780-250MG
APOSOR3780-1GEA 46.2 GBP
APOSOR3780-1G APOSOR3780-250MG
4-(Tributylstannyl)pyridine
4-(Tributylstannyl)pyridine
Formula: C₁₇H₃₁NSn
Storage Temperature: Ambient
MDL Number: MFCD01319071
CAS Number: 124252-41-1
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR