Isobutyraldehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Methylpropanal, 2-Methylpropionaldehyde

OR322092-2.5L OR322092-500ML
APOSOR322092-2.5LEA 90.7 GBP
APOSOR322092-2.5L APOSOR322092-500ML
Isobutyraldehyde 98%
Isobutyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 63 °C
Melting Pt: –65 °C
Density: 0.79 g/cm³ (25 °C)
Flash Pt: –18.99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006980
CAS Number: 78-84-2
UN: 2045
ADR: 3 II
Merck Index: 13,05171

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR