cis-3-(Benzyloxy)cyclobutanamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR317136-1G OR317136-250MG
APOSOR317136-1GEA 525 GBP
APOSOR317136-1G APOSOR317136-250MG
cis-3-(Benzyloxy)cyclobutanamine
cis-3-(Benzyloxy)cyclobutanamine
Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660354
CAS Number: 206660-72-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR