1,4,8,11-Tetraazacyclotetradecane 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
CYCLAM
OR3122-1G
OR3122-5G
APOSOR3122-1GEA
16.8
GBP
APOSOR3122-1G
APOSOR3122-5G
1,4,8,11-Tetraazacyclotetradecane 98%
1,4,8,11-Tetraazacyclotetradecane
Formula:
C₁₀H₂₄N₄ Melting Pt: 184…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005105 CAS Number: 295-37-4 EINECS: 206-039-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...