5-(Chloromethyl)-3-(3-thienyl)-1,2,4-oxadiazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR310756-1G OR310756-5G
APOSOR310756-1GEA 214 GBP
APOSOR310756-1G APOSOR310756-5G
5-(Chloromethyl)-3-(3-thienyl)-1,2,4-oxadiazole
5-(Chloromethyl)-3-(3-thienyl)-1,2,4-oxadiazole
Formula: C₇H₅ClN₂OS
Storage Temperature: Ambient
MDL Number: MFCD07343884
CAS Number: 869709-84-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR