Chloroiodomethane 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OR30688-25G
OR30688-100G
OR30688-500G
APOSOR30688-25GEA
15.8
GBP
APOSOR30688-25G
APOSOR30688-100G
APOSOR30688-500G
Chloroiodomethane 98%
Chloroiodomethane
Formula:
CH₂ClI Boiling Pt: 108…109 °C Density: 2.422 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001078 CAS Number: 593-71-5 EINECS: 209-804-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...