Piperazine-1-carboxamidinium acetate
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR300536-1G
OR300536-5G
APOSOR300536-1GEA
94.9
GBP
APOSOR300536-1G
APOSOR300536-5G
Piperazine-1-carboxamidinium acetate
Piperazine-1-carboxamidinium acetate
Formula:
C₇H₁₆N₄O₂ MW: 188.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568300 CAS Number: 60729-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...