N,N-Dimethylformamide dimethyl acetal
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
OR28611-100G
OR28611-1KG
OR28611-500G
APOSOR28611-100GEA
15.8
GBP
APOSOR28611-100G
APOSOR28611-1KG
APOSOR28611-500G
N,N-Dimethylformamide dimethyl acetal
N,N-Dimethylformamide dimethyl acetal
Formula:
C₅H₁₃NO₂ Boiling Pt: 102…104 °C Density: 0.897 g/cm³ (25 °C) Flash Pt: 5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008482 CAS Number: 4637-24-5 EINECS: 225-063-3 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...