1-Ethoxy-1-(tributylstannyl)ethene 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Tributyl(1-ethoxyvinyl)tin, Tri-n-butyl(1-ethoxyvinyl)tin, 1-Ethoxyvinyltri-n-butyltin

OR2395-25G OR2395-5G OR2395-100G
APOSOR2395-25GEA 32.6 GBP
APOSOR2395-25G APOSOR2395-5G APOSOR2395-100G
1-Ethoxy-1-(tributylstannyl)ethene 95%
1-Ethoxy-1-(tributylstannyl)ethene
Formula: C₁₆H₃₄OSn
Boiling Pt: 85…86 °C/0.1mmHg
Density: 1.069 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00010240
CAS Number: 97674-02-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR