3-Pyridylmethanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol

OR23061-25G OR23061-100G OR23061-500G
APOSOR23061-25GEA 16.8 GBP
APOSOR23061-25G APOSOR23061-100G APOSOR23061-500G
3-Pyridylmethanol
3-Pyridylmethanol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 259 °C
Melting Pt: –7 °C
Density: 1.134 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR