Tributyltin chloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR2288-250G OR2288-50G OR2288-5G
APOSOR2288-250GEA 99 GBP
APOSOR2288-250G APOSOR2288-50G APOSOR2288-5G
Tributyltin chloride 95%
Tributyltin chloride
Formula: C₁₂H₂₇ClSn
Boiling Pt: 171…173 °C/25mmHg
Melting Pt: –19 °C
Density: 1.186 g/cm³ (25 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00000521
CAS Number: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR