Tecnazene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tecnazene, 2,3,5,6-Tetrachloronitrobenzene

OR22833-25G OR22833-500G
APOSOR22833-25GEA 83.9 GBP
APOSOR22833-25G APOSOR22833-500G
Tecnazene
Tecnazene
Formula: C₆HCl₄NO₂
MW: 260.89 g/mol
Boiling Pt: 304
Melting Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00007066
CAS Number: 117-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR