Acetyl bromide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR1943-25G OR1943-500G
APOSOR1943-25GEA 42 GBP
APOSOR1943-25G APOSOR1943-500G
Acetyl bromide 98%
Acetyl bromide
Formula: C₂H₃BrO
Boiling Pt: 75…77 °C
Melting Pt: –96 °C
Density: 1.663 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000114
CAS Number: 506-96-7
EINECS: 208-061-7
UN: 1716
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR