Tributylallyltin

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Allyltributylstannane, Allyltributyltin, Tributylallylstannane, Tributyl-2-propenylstannane, Allyltri-n-butyltin, Tri-n-butyl-2-propenylstannane, Allyltri-n-butylstannane

OR16616-100G OR16616-25G
APOSOR16616-100GEA 258 GBP
APOSOR16616-100G APOSOR16616-25G
Tributylallyltin
Tributylallyltin
Formula: C₁₅H₃₂Sn
Boiling Pt: 88…92 °C/0.2mmHg
Density: 1.068 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR