Triethyl orthoformate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR16612-1L OR16612-2.5L
APOSOR16612-1LEA 137 GBP
APOSOR16612-1L APOSOR16612-2.5L
Triethyl orthoformate 98%
Triethyl orthoformate
Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 143…147 °C
Melting Pt: –61 °C
Density: 0.89 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
EINECS: 204-550-4
UN: 2524
ADR: 3 III
Merck Index: 12,07012

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR