2-(Tributylstannyl)thiazole 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15628-1G OR15628-5G
APOSOR15628-1GEA 47.3 GBP
APOSOR15628-1G APOSOR15628-5G
2-(Tributylstannyl)thiazole 95+%
2-(Tributylstannyl)thiazole
Formula: C₁₅H₂₉NSSn
MW: 374.17 g/mol
Boiling Pt: 307…309 °C
Density: 1.19 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319057
CAS Number: 121359-48-6
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR