4-(Tributylstannyl)pyridazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15627-1G OR15627-250MG OR15627-500MG
APOSOR15627-1GEA 426 GBP
APOSOR15627-1G APOSOR15627-250MG APOSOR15627-500MG
4-(Tributylstannyl)pyridazine
4-(Tributylstannyl)pyridazine
Formula: C₁₆H₃₀N₂Sn
Storage Temperature: Refrigerator
MDL Number: MFCD07787369
CAS Number: 194865-89-9
UN: 2788
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR