2-Methyl-6-(tributylstannyl)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15619-1G OR15619-250MG
APOSOR15619-1GEA 17.9 GBP
APOSOR15619-1G APOSOR15619-250MG
2-Methyl-6-(tributylstannyl)pyridine
2-Methyl-6-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NSn
Density: 1.134 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787401
CAS Number: 259807-95-9
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR