4-Methyl-2-(tributylstannyl)pyridine 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
OR15617-1G
OR15617-250MG
APOSOR15617-1GEA
168
GBP
APOSOR15617-1G
APOSOR15617-250MG
4-Methyl-2-(tributylstannyl)pyridine 97%
4-Methyl-2-(tributylstannyl)pyridine
Formula:
C₁₈H₃₃NSn Storage Temperature: Ambient |
MDL Number:
MFCD07787403 CAS Number: 301652-23-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...