3,5-Dimethyl-4-(tributylstannyl)isoxazole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15592-1G OR15592-250MG
APOSOR15592-1GEA 247 GBP
APOSOR15592-1G APOSOR15592-250MG
3,5-Dimethyl-4-(tributylstannyl)isoxazole 95%
3,5-Dimethyl-4-(tributylstannyl)isoxazole
Formula: C₁₇H₃₃NOSn
Storage Temperature: Ambient
MDL Number: MFCD01318973
CAS Number: 136295-80-2
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR