1-Methyl-2-(tributylstannyl)imidazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15578-1G OR15578-250MG
APOSOR15578-1GEA 174 GBP
APOSOR15578-1G APOSOR15578-250MG
1-Methyl-2-(tributylstannyl)imidazole
1-Methyl-2-(tributylstannyl)imidazole
Formula: C₁₆H₃₂N₂Sn
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319030
CAS Number: 105494-69-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR