5-(Tributylstannyl)thiazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15547-1G OR15547-250MG OR15547-5G
APOSOR15547-1GEA 75.6 GBP
APOSOR15547-1G APOSOR15547-250MG APOSOR15547-5G
5-(Tributylstannyl)thiazole
5-(Tributylstannyl)thiazole
Formula: C₁₅H₂₉NSSn
Storage Temperature: Refrigerator
MDL Number: MFCD01319074
CAS Number: 157025-33-7
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR