4-Methyl-2-(tributylstannyl)thiazole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR15544-1G OR15544-250MG
APOSOR15544-1GEA 279 GBP
APOSOR15544-1G APOSOR15544-250MG
4-Methyl-2-(tributylstannyl)thiazole 95%
4-Methyl-2-(tributylstannyl)thiazole
Formula: C₁₆H₃₁NSSn
Storage Temperature: Refrigerator
MDL Number: MFCD09025808
CAS Number: 251635-59-3
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR