2-Bromobiphenyl (PBB No. 1)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromobiphenyl, PBB No. 1, 2-Bromodiphenyl

OR13698-100G OR13698-25G OR13698-5G
APOSOR13698-100GEA 108 GBP
APOSOR13698-100G APOSOR13698-25G APOSOR13698-5G
2-Bromobiphenyl (PBB No. 1)
2-Bromobiphenyl (PBB No. 1)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 297…298 °C
Melting Pt: 1.5…2 °C
Density: 1.352 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000065
CAS Number: 2052-07-5
EINECS: 218-141-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR